Learn More
Medchemexpress LLC HY-12111 10mg , BMS-564929 CAS:627530-84-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1211110MG
Encompass_Preferred
HY-12111 10mg BMS-564929 CAS: 627530-84-1 BMS-564929 is an androgen receptor (AR) agonist, binds to androgen receptor (AR) with a Ki of 2.11±0.16 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0605