Learn More
Selleck Chemical LLC BMS-582949 1mg 623152-17-0 PS540446
SDP
Catalog No. 501365680
Encompass_Preferred
BMS-582949 (PS540446) is a potent and selective p38 mitogen-activated protein kinase (p38 MAPK) inhibitor with IC50 of 13nM,inhibiting both p38 kinase activity and activation of p38. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-5680 Supplier Selleck Chemical LLC Supplier No. S81241MG
BMS-582949 (PS540446) is a potent and selective p38 mitogen-activated protein kinase (p38 MAPK) inhibitor with IC50 of 13nM,inhibiting both p38 kinase activity and activation of p38. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.