Learn More
Medchemexpress LLC HY-14305A 5mg Medchemexpress, BMS-582949 (hydrochloride) CAS:912806-16-7 Purity:>98%
SDP
Catalog No. 502030527
Encompass_Preferred
Medchemexpress, HY-14305A 5mg BMS-582949 (hydrochloride) CAS:912806-16-7 BMS-582949 hydrochloride is an orally active and highly selective p38α MAPK inhibitor, with an IC50 of 13 nM. BMS-582949 hydrochloride displays a significantly improved pharmacokinetic profile and is effective in inflammatory disease. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-0527 Supplier Medchemexpress LLC Supplier No. HY14305A5MG
Medchemexpress, HY-14305A 5mg BMS-582949 (hydrochloride) CAS:912806-16-7 BMS-582949 hydrochloride is an orally active and highly selective p38α MAPK inhibitor, with an IC50 of 13 nM. BMS-582949 hydrochloride displays a significantly improved pharmacokinetic profile and is effective in inflammatory disease. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.