Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC BMS-593214 25mg | 1004551-40-9 | 505.56 g·mol-1 | C31H27N3O4 | 25 MG
SDP

Catalog No. 5000229586
Encompass_Preferred

BMS-593214 is a research-grade small-molecule inhibitor of activated coagulation factor VII (FVIIa). It acts as a direct competitive inhibitor of human FVIIa, demonstrates antithrombotic activity in preclinical models, and is supplied as a purified solid for laboratory research use only.

  • High purity (99.90%) suitable for biochemical and pharmacology assays.
  • Soluble in DMSO at high concentration; ultrasonic agitation may improve dissolution.
  • Recommended storage conditions available to maintain stability as a powder and in solution.
  • Provided in a 25 mg pack convenient for small-scale experiments.
  • Intended for research use only; not for human or veterinary use.

Catalog No. 50-002-29586 Supplier Medchemexpress LLC Supplier No. HY11451125MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

BMS-593214 is a research-grade small-molecule inhibitor of activated coagulation factor VII (FVIIa). It acts as a direct competitive inhibitor of human FVIIa, demonstrates antithrombotic activity in preclinical models, and is supplied as a purified solid for laboratory research use only.

  • High purity (99.90%) suitable for biochemical and pharmacology assays.
  • Soluble in DMSO at high concentration; ultrasonic agitation may improve dissolution.
  • Recommended storage conditions available to maintain stability as a powder and in solution.
  • Provided in a 25 mg pack convenient for small-scale experiments.
  • Intended for research use only; not for human or veterinary use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.