Learn More
Selleck Chemical LLC AC480 (BMS-599626) 5mg 714971-09-2
SDP
Catalog No. 507792
Encompass_Preferred
AC480 (BMS-599626) is a selective and efficacious inhibitor of HER1 and HER2 with IC50 of 20 nM and 30 nM, ~8-fold less potent to HER4, >100-fold to VEGFR2, c-Kit, Lck, MET etc. Phase 1. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-779-2 Supplier Selleck Chemical LLC Supplier No. S10565MG
AC480 (BMS-599626) is a selective and efficacious inhibitor of HER1 and HER2 with IC50 of 20 nM and 30 nM, ~8-fold less potent to HER4, >100-fold to VEGFR2, c-Kit, Lck, MET etc. Phase 1. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.