Learn More
Medchemexpress LLC HY-12010 5mg Medchemexpress, BMS-599626 (Hydrochloride) CAS:873837-23-1 Purity:>98%
SDP
Catalog No. 501969576
Encompass_Preferred
Medchemexpress, HY-12010 5mg BMS-599626 (HCl) CAS:873837-23-1 BMS-599626 Hydrochloride (AC480 Hydrochloride) is a selective and orally bioavailable HER1 and HER2 inhibitor, with IC50s of 20 and 30 nM, respectively. BMS-599626 Hydrochloride displays ~8-fold less potent to HER4 (IC50=190 nM), >100-fold to VEGFR2, c-Kit, Lck, MEK. BMS-599626 Hydrochloride inhibits tumor cell proliferation, and has potential to increase tumor response to radiotherapy. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9576 Supplier Medchemexpress LLC Supplier No. HY120105MG
Medchemexpress, HY-12010 5mg BMS-599626 (HCl) CAS:873837-23-1 BMS-599626 Hydrochloride (AC480 Hydrochloride) is a selective and orally bioavailable HER1 and HER2 inhibitor, with IC50s of 20 and 30 nM, respectively. BMS-599626 Hydrochloride displays ~8-fold less potent to HER4 (IC50=190 nM), >100-fold to VEGFR2, c-Kit, Lck, MEK. BMS-599626 Hydrochloride inhibits tumor cell proliferation, and has potential to increase tumor response to radiotherapy. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.