Learn More
Apexbio Technology LLC BMS-626529 701213-36-7 200mg
SDP
Catalog No. 5000063602
Encompass_Preferred
BMS-626529 is a small molecule inhibitor designed to block HIV-1 attachment to host cells Its mechanism involves binding to the viral envelope protein gp120 thereby hindering gp120-mediated interactions with CD4 receptors on CD4 T cells and preventing initial viral entry In cell-based assays BMS-626529 shows strain-specific antiviral activity with IC50 values mostly below 10 nM and notably low picomolar IC50 against highly susceptible HIV-1 isolates This molecule serves as a research tool to investigate HIV-1 entry inhibition and virus-host interaction mechanisms in biomedical studies
Catalog No. 50-000-63602 Supplier Apexbio Technology LLC Supplier No. A3253200
BMS-626529 is a small molecule inhibitor designed to block HIV-1 attachment to host cells Its mechanism involves binding to the viral envelope protein gp120 thereby hindering gp120-mediated interactions with CD4 receptors on CD4 T cells and preventing initial viral entry In cell-based assays BMS-626529 shows strain-specific antiviral activity with IC50 values mostly below 10 nM and notably low picomolar IC50 against highly susceptible HIV-1 isolates This molecule serves as a research tool to investigate HIV-1 entry inhibition and virus-host interaction mechanisms in biomedical studies