Learn More
Medchemexpress LLC HY-10678 5mg Medchemexpress, BMS-687453 CAS:1000998-59-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY106785MG
Encompass_Preferred
Medchemexpress, HY-10678 5mg BMS-687453 CAS:1000998-59-3 BMS-687453 is a potent and selective PPARα agonist, with an EC50 and IC50 of 10 nM and 260 nM for human PPARα and 4100 nM and >15000 nM for PPARγ in PPAR-GAL4 transactivation assays. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8353