Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (3R,4R)-4-amino-1-[[4-(3-methoxyanilino)pyrrolo[2,1-f]triazin-5-yl]methyl]piperidin-3-ol | 859853-30-8 | MFCD18633295 | 99.7% | 368.43 g/mol | C19H24N6O2 | 2 MG
SDP

Catalog No. 502028728
Encompass_Preferred

BMS-690514 is a small-molecule, orally active tyrosine kinase inhibitor that targets epidermal growth factor receptors (EGFR/HER family) and vascular endothelial growth factor receptors (VEGFR). It has been investigated in preclinical and clinical studies for treatment of solid tumors and is supplied for research use only.

  • Potent inhibition of EGFR, HER2, and HER4 (IC50 ≈5, 20, 60 nM).
  • Activity against VEGFR family members and additional kinases.
  • Orally active with documented preclinical bioavailability in multiple species.
  • High purity suitable for research (99.68%).
  • Molecular formula C19H24N6O2; molecular weight 368.43 g/mol.
  • Intended for laboratory research and preclinical studies only.

Catalog No. 50-202-8728 Supplier Medchemexpress LLC Supplier No. HY103332MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

BMS-690514 is a small-molecule, orally active tyrosine kinase inhibitor that targets epidermal growth factor receptors (EGFR/HER family) and vascular endothelial growth factor receptors (VEGFR). It has been investigated in preclinical and clinical studies for treatment of solid tumors and is supplied for research use only.

  • Potent inhibition of EGFR, HER2, and HER4 (IC50 ≈5, 20, 60 nM).
  • Activity against VEGFR family members and additional kinases.
  • Orally active with documented preclinical bioavailability in multiple species.
  • High purity suitable for research (99.68%).
  • Molecular formula C19H24N6O2; molecular weight 368.43 g/mol.
  • Intended for laboratory research and preclinical studies only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.