Learn More
Selleck Chemical LLC Avagacestat (BMS-708163) 50mg 1146699-66-2
SDP
Catalog No. 507296
Encompass_Preferred
Avagacestat (BMS-708163) is a potent, selective, orally bioavailable ?-secretase inhibitor of A?40 and A?42 with IC50 of 0.3 nM and 0.27 nM, demonstrating a 193-fold selectivity against Notch. Phase 2. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-729-6 Supplier Selleck Chemical LLC Supplier No. S126250MG
Avagacestat (BMS-708163) is a potent, selective, orally bioavailable ?-secretase inhibitor of A?40 and A?42 with IC50 of 0.3 nM and 0.27 nM, demonstrating a 193-fold selectivity against Notch. Phase 2. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.