Learn More
Medchemexpress LLC HY-10200 10mg Medchemexpress, BMS-754807 CAS:1001350-96-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1020010MG
Encompass_Preferred
Medchemexpress, HY-10200 10mg BMS-754807 CAS:1001350-96-4 BMS-754807 is a potent and reversible IGF-1R/IR inhibitor (IC50=1.8 and 1.7 nM, respectively; Ki=<2 nM for both). BMS-754807 also shows potent activities against Met, RON, TrkA, TrkB, AurA, and AurB with IC50 values of 6, 44, 7, 4, 9, and 25 nM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-8695