Learn More
Selleck Chemical LLC BMS-754807 5mg 1001350-96-4
SDP
Catalog No. NC0585502
Encompass_Preferred
BMS-754807 is a potent and reversible inhibitor of IGF-1R/InsR with IC50 of 1.8 nM/1.7 nM in cell-free assays, less potent to Met (c-Met), Aurora A/B, TrkA/B and Ron, and shows little activity to Flt3, Lck, MK2, PKA, PKC etc. Phase 2. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC0585502 Supplier Selleck Chemical LLC Supplier No. S11245MG
BMS-754807 is a potent and reversible inhibitor of IGF-1R/InsR with IC50 of 1.8 nM/1.7 nM in cell-free assays, less potent to Met (c-Met), Aurora A/B, TrkA/B and Ron, and shows little activity to Flt3, Lck, MK2, PKA, PKC etc. Phase 2. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.