Learn More
Medchemexpress LLC HY-10200 100mg , BMS-754807 CAS:1001350-96-4 Purity:98%
SDP
Catalog No. 501492996
Encompass_Preferred
Medchemexpress, HY-10200 100mg BMS-754807 CAS:1001350-96-4 Formula:C23H24FN9O IC50: 1.7 nM (insulin receptor), 1.8 nM (IGF-1R), 4 nM (TrkB), 6 nM (Met), 7 nM (TrkA), 9 nM (AurA), 25 nM (AurB), 44 nM (RON) Purity:98% BMS-754807 is a potent and reversible inhibitor of the insulin-like growth factor 1 receptor (IGF-1R) /insulin receptor family kinases (IR) with IC 50 of 1.8 and 1.7 nM, respectively and K i of <2 nM for both, and also shows potent activities against Met, RON, TrkA, TrkB, AurA, and AurB with IC 50 values of 6, 44, 7, 4, 9, and 25 nM, respectively. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-2996 Supplier Medchemexpress LLC Supplier No. HY10200100MG
Medchemexpress, HY-10200 100mg BMS-754807 CAS:1001350-96-4 Formula:C23H24FN9O IC50: 1.7 nM (insulin receptor), 1.8 nM (IGF-1R), 4 nM (TrkB), 6 nM (Met), 7 nM (TrkA), 9 nM (AurA), 25 nM (AurB), 44 nM (RON) Purity:98% BMS-754807 is a potent and reversible inhibitor of the insulin-like growth factor 1 receptor (IGF-1R) /insulin receptor family kinases (IR) with IC 50 of 1.8 and 1.7 nM, respectively and K i of <2 nM for both, and also shows potent activities against Met, RON, TrkA, TrkB, AurA, and AurB with IC 50 values of 6, 44, 7, 4, 9, and 25 nM, respectively. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.