Learn More
Apexbio Technology LLC BMS-777607 1025720-94-8 10mM (in 1mL DMSO)
SDP
Catalog No. 5000062442
Encompass_Preferred
BMS-777607 (CAS 1025720-94-8) is an orally bioavailable ATP-competitive inhibitor targeting MET kinase and related receptor tyrosine kinases It selectively inhibits members of the MET family including RON MET Tyro-3 and Axl with IC50 values of 1 8 nmol/L 3 9 nmol/L 4 3 nmol/L and 1 1 nmol/L respectively At higher doses it also inhibits additional tyrosine kinases such as Mer Flt-3 Aurora B Lck and VEGFR2 BMS-777607 has been shown to suppress c-Met autophosphorylation (IC50 20 nmol/L) thereby impairing tumor xenograft growth and metastatic phenotypes supporting its utility in oncological research models exploring MET-driven tumorigenesis and cancer progression
Catalog No. 50-000-62442 Supplier Apexbio Technology LLC Supplier No. A57035.1
BMS-777607 (CAS 1025720-94-8) is an orally bioavailable ATP-competitive inhibitor targeting MET kinase and related receptor tyrosine kinases It selectively inhibits members of the MET family including RON MET Tyro-3 and Axl with IC50 values of 1 8 nmol/L 3 9 nmol/L 4 3 nmol/L and 1 1 nmol/L respectively At higher doses it also inhibits additional tyrosine kinases such as Mer Flt-3 Aurora B Lck and VEGFR2 BMS-777607 has been shown to suppress c-Met autophosphorylation (IC50 20 nmol/L) thereby impairing tumor xenograft growth and metastatic phenotypes supporting its utility in oncological research models exploring MET-driven tumorigenesis and cancer progression