Learn More
Selleck Chemical LLC BMS-777607 10mM/1mL 1025720-94-8
SDP
Catalog No. 501361389
Encompass_Preferred
BMS-777607 is a Met-related inhibitor for c-Met, Axl, Ron and Tyro3 with IC50 of 3.9 nM, 1.1 nM, 1.8 nM and 4.3 nM in cell-free assays, 40-fold more selective for Met-related targets versus Lck, VEGFR-2, and TrkA/B, and more than 500-fold greater selectivity versus all other receptor and non receptor kinases. Phase 1/2. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-1389 Supplier Selleck Chemical LLC Supplier No. S156110MM/1ML
BMS-777607 is a Met-related inhibitor for c-Met, Axl, Ron and Tyro3 with IC50 of 3.9 nM, 1.1 nM, 1.8 nM and 4.3 nM in cell-free assays, 40-fold more selective for Met-related targets versus Lck, VEGFR-2, and TrkA/B, and more than 500-fold greater selectivity versus all other receptor and non receptor kinases. Phase 1/2. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.