Learn More
Medchemexpress LLC HY-12076 50mg , BMS 777607 CAS:1025720-94-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1207650MG
Encompass_Preferred
HY-12076 50mg BMS 777607 CAS: 1025720-94-8 BMS 777607 (BMS 817378) is a Met-related inhibitor for c-Met, Axl, Ron and Tyro3 with IC50s of 3.9 nM, 1.1 nM, 1.8 nM and 4.3 nM, respectively, and 40-fold more selective for Met-related targets than Lck, VEGFR-2, and TrkA/B, with more than 500-fold greater selectivity versus all other receptor and non receptor kinases. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0600