Learn More
TARGETMOL CHEMICALS INC BMS-794833 10MG
SDP
Catalog No. 502580823
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. BMS-794833 is a potent ATP competitive inhibitor of Met/VEGFR2; a prodrug of BMS-817378. purity: 99%
Catalog No. 50-258-0823 Supplier TARGETMOL CHEMICALS INC Supplier No. T241910MG
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. BMS-794833 is a potent ATP competitive inhibitor of Met/VEGFR2; a prodrug of BMS-817378. purity: 99%