Learn More
Medchemexpress LLC HY-10497 5mg Medchemexpress, BMS-794833 CAS:1174046-72-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY104975MG
Encompass_Preferred
Medchemexpress, HY-10497 5mg BMS-794833 CAS:1174046-72-0 BMS-794833 is a VEGFR2 and Met inhibitor extracted from patent WO2009094417, compound example 1; has IC50s of 15 and 1.7 nM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8238