Learn More
Medchemexpress LLC BMS-8 10mg | 1675201-90-7 | 494.42 g·mol⁻¹ | C27H28BrNO3 | 10 MG
SDP
Catalog No. 5000228831
Encompass_Preferred
BMS-8 is a small-molecule inhibitor of the PD-1/PD-L1 protein-protein interaction used in biochemical and cell-based studies of immune checkpoint modulation. It binds directly to PD-L1, promotes PD-L1 homodimer formation, and inhibits PD-1/PD-L1 binding with a reported IC50 of 7.2 μM.
  • Small-molecule PD-1/PD-L1 inhibitor for research applications.
  • Direct PD-L1 binder that induces homodimer formation.
  • Reported IC50 of 7.2 μM against PD-1/PD-L1 interaction.
  • High purity suitable for biochemical assays.
  • Available as solid milligram quantities and as DMSO solutions for assay use.
Catalog No. 50-002-28831 Supplier Medchemexpress LLC Supplier No. HY11627410MG
Add to Cart
Add to Cart
BMS-8 is a small-molecule inhibitor of the PD-1/PD-L1 protein-protein interaction used in biochemical and cell-based studies of immune checkpoint modulation. It binds directly to PD-L1, promotes PD-L1 homodimer formation, and inhibits PD-1/PD-L1 binding with a reported IC50 of 7.2 μM.
  • Small-molecule PD-1/PD-L1 inhibitor for research applications.
  • Direct PD-L1 binder that induces homodimer formation.
  • Reported IC50 of 7.2 μM against PD-1/PD-L1 interaction.
  • High purity suitable for biochemical assays.
  • Available as solid milligram quantities and as DMSO solutions for assay use.