Learn More
TARGETMOL CHEMICALS INC BMS-813160 5MG
SDP
Catalog No. 502576747
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. BMS-813160 is the first dual CCR2/CCR5 antagonist to enter Clinical development for cardiovascular. purity: 99%
Catalog No. 50-257-6747 Supplier TARGETMOL CHEMICALS INC Supplier No. T45845MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. BMS-813160 is the first dual CCR2/CCR5 antagonist to enter Clinical development for cardiovascular. purity: 99%