Learn More
Medchemexpress LLC HY-101930 5mg Medchemexpress, BMS-816336 CAS:1009583-20-3 Purity:>98%
SDP
Catalog No. 501967921
Encompass_Preferred
Medchemexpress, HY-101930 5mg BMS-816336 CAS:1009583-20-3 BMS-816336 is a novel, potent and orally bioavailable inhibitor against human 11β-hydroxysteroid dehydrogenase type 1 (11β-HSD1) enzyme with an IC50 of 3.0 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-7921 Supplier Medchemexpress LLC Supplier No. HY1019305MG
Medchemexpress, HY-101930 5mg BMS-816336 CAS:1009583-20-3 BMS-816336 is a novel, potent and orally bioavailable inhibitor against human 11β-hydroxysteroid dehydrogenase type 1 (11β-HSD1) enzyme with an IC50 of 3.0 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.