Learn More
Medchemexpress LLC HY-15670 5mg Medchemexpress, BMS-906024 CAS:1401066-79-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY156705MG
Encompass_Preferred
Medchemexpress, HY-15670 5mg BMS-906024 CAS:1401066-79-2 BMS-906024 is an orally active and selective γ-secretase (gamma secretase) inhibitor. BMS-906024 is a potent pan-Notch receptors inhibitor with IC50s of 1.6 nM, 0.7 nM, 3.4 nM, and 2.9 nM for Notch1, -2, -3, and -4 receptors, respectively. BMS-906024 demonstrates broad-spectrum antineoplastic activity. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1766