Learn More
Medchemexpress LLC BMS-911172 100mg | 1644248-18-9 | 339.34 | C16H19F2N3O3 | 100 MG
SDP
Catalog No. 5000229253
Encompass_Preferred
BMS-911172 is a small-molecule inhibitor of adaptor-associated kinase 1 (AAK1) with a reported IC50 of 35 nM; it is supplied for laboratory research use only.
  • Inhibits AAK1 with IC50 of 35 nM
  • Reported purity 99.49%
  • Available in multiple solid sizes and 10 mM DMSO solutions
  • Suitable for in vitro and preclinical research; not for human use
  • Molecular weight 339.34 g/mol; formula C16H19F2N3O3
  • Certificate of analysis and safety data sheet available
Catalog No. 50-002-29253 Supplier Medchemexpress LLC Supplier No. HY115868100MG
BMS-911172 is a small-molecule inhibitor of adaptor-associated kinase 1 (AAK1) with a reported IC50 of 35 nM; it is supplied for laboratory research use only.
  • Inhibits AAK1 with IC50 of 35 nM
  • Reported purity 99.49%
  • Available in multiple solid sizes and 10 mM DMSO solutions
  • Suitable for in vitro and preclinical research; not for human use
  • Molecular weight 339.34 g/mol; formula C16H19F2N3O3
  • Certificate of analysis and safety data sheet available