Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC BMS-911172 10mg | 1644248-18-9 | 339.34 | C16H19F2N3O3 | 10 MG
SDP

Catalog No. 5000226692
Encompass_Preferred

BMS-911172 is a small-molecule inhibitor of adaptor-associated kinase 1 (AAK1) provided for laboratory research. It has reported nanomolar potency and is offered at high purity for biochemical and cellular studies.

  • Selective AAK1 inhibition with nanomolar potency (IC50 ≈ 35 nM).
  • High reported purity (≈99.5%).
  • Molecular weight 339.34; formula C16H19F2N3O3.
  • Soluble in DMSO; solution format available.
  • Available in small-scale quantities suitable for research workflows.
  • For research use only; not for clinical or human use.

Catalog No. 50-002-26692 Supplier Medchemexpress LLC Supplier No. HY11586810MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

BMS-911172 is a small-molecule inhibitor of adaptor-associated kinase 1 (AAK1) provided for laboratory research. It has reported nanomolar potency and is offered at high purity for biochemical and cellular studies.

  • Selective AAK1 inhibition with nanomolar potency (IC50 ≈ 35 nM).
  • High reported purity (≈99.5%).
  • Molecular weight 339.34; formula C16H19F2N3O3.
  • Soluble in DMSO; solution format available.
  • Available in small-scale quantities suitable for research workflows.
  • For research use only; not for clinical or human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.