Learn More
Medchemexpress LLC BMS-911172 25mg | 1644248-18-9 | 339.34 g/mol | C16H19F2N3O3 | 25 MG
SDP
Catalog No. 5000308265
Encompass_Preferred
BMS-911172 is a small-molecule, brain-penetrant inhibitor of adaptor protein 2-associated kinase 1 (AAK1) used as a research compound to investigate AAK1-mediated signaling and neuropathic pain mechanisms. Chemical formula C16H19F2N3O3; molecular weight 339.34 g/mol; reported AAK1 activity in the low-nanomolar range.
  • Potent AAK1 inhibition with low-nanomolar activity.
  • Brain-penetrant small molecule suitable for in vivo models.
  • High purity appropriate for biochemical and cellular assays.
  • Available in small research quantities for laboratory studies.
Catalog No. 50-003-08265 Supplier Medchemexpress LLC Supplier No. HY11586825MG
BMS-911172 is a small-molecule, brain-penetrant inhibitor of adaptor protein 2-associated kinase 1 (AAK1) used as a research compound to investigate AAK1-mediated signaling and neuropathic pain mechanisms. Chemical formula C16H19F2N3O3; molecular weight 339.34 g/mol; reported AAK1 activity in the low-nanomolar range.
  • Potent AAK1 inhibition with low-nanomolar activity.
  • Brain-penetrant small molecule suitable for in vivo models.
  • High purity appropriate for biochemical and cellular assays.
  • Available in small research quantities for laboratory studies.