Learn More
Medchemexpress LLC BMS-911172 50mg | 1644248-18-9 | 339.34 g/mol | C16H19F2N3O3 | 50 MG
SDP
Catalog No. 5000228095
Encompass_Preferred
BMS-911172 is a small-molecule, brain-penetrant inhibitor of adaptor protein 2-associated kinase 1 (AAK1) supplied as a 50 mg solid for research use only. It was developed as a tool compound to validate AAK1 as a target for neuropathic pain; reported potency varies across sources.
  • Inhibits AAK1 with reported IC50 in the low nanomolar range (reported values 12-35 nM).
  • Brain-penetrant small molecule suitable for in vitro and in vivo studies.
  • Supplied as a solid at 50 mg quantity.
  • High supplier-reported purity (typically ≥99%).
  • Provided for research use only; not for human therapeutic use.
Catalog No. 50-002-28095 Supplier Medchemexpress LLC Supplier No. HY11586850MG
BMS-911172 is a small-molecule, brain-penetrant inhibitor of adaptor protein 2-associated kinase 1 (AAK1) supplied as a 50 mg solid for research use only. It was developed as a tool compound to validate AAK1 as a target for neuropathic pain; reported potency varies across sources.
  • Inhibits AAK1 with reported IC50 in the low nanomolar range (reported values 12-35 nM).
  • Brain-penetrant small molecule suitable for in vitro and in vivo studies.
  • Supplied as a solid at 50 mg quantity.
  • High supplier-reported purity (typically ≥99%).
  • Provided for research use only; not for human therapeutic use.