Learn More
Medchemexpress LLC BMS-911172 5mg | 1644248-18-9 | 5 MG
SDP
Catalog No. 5000308034
Encompass_Preferred
BMS-911172 is a small-molecule inhibitor of adaptor protein 2-associated kinase 1 (AAK1) supplied as a solid for laboratory research. It is brain-penetrant and reported to have low-nanomolar potency in biochemical and cellular assays (reported IC50 values ≈12-35 nM). For research use only.
  • Potent AAK1 inhibitor with low-nanomolar activity.
  • Brain-penetrant small molecule suitable for CNS-targeted studies.
  • High purity (~99.5%) solid form for analytical and biological work.
  • Supplied in a small quantity suitable for pilot or mechanistic studies.
  • Intended for research use only; not for human or clinical use.
Catalog No. 50-003-08034 Supplier Medchemexpress LLC Supplier No. HY1158685MG
BMS-911172 is a small-molecule inhibitor of adaptor protein 2-associated kinase 1 (AAK1) supplied as a solid for laboratory research. It is brain-penetrant and reported to have low-nanomolar potency in biochemical and cellular assays (reported IC50 values ≈12-35 nM). For research use only.
  • Potent AAK1 inhibitor with low-nanomolar activity.
  • Brain-penetrant small molecule suitable for CNS-targeted studies.
  • High purity (~99.5%) solid form for analytical and biological work.
  • Supplied in a small quantity suitable for pilot or mechanistic studies.
  • Intended for research use only; not for human or clinical use.