Learn More
Medchemexpress LLC HY-15270 10mg Medchemexpress, BMS-911543 CAS:1271022-90-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1527010MG
Encompass_Preferred
Medchemexpress, HY-15270 10mg BMS-911543 CAS:1271022-90-2 BMS-911543 is a selective JAK2 inhibitor, with IC50s of 1.1 nM, less selective at JAK1, JAK3 and TYK2 (IC50, 75, 360, 66 nM, respectively). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9110