Learn More
TARGETMOL CHEMICALS INC BMS-929075 5MG
SDP
Catalog No. 502580667
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. BMS-929075 is an orally active HCV NS5B replicase (HCV NS5B replicase) palm site variant inhibitor with potency high oral bioavailability and pharmacokinetic parameters.BMS-929075 shows cytotoxicity. purity: 97%
Catalog No. 50-258-0667 Supplier TARGETMOL CHEMICALS INC Supplier No. T268635MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. BMS-929075 is an orally active HCV NS5B replicase (HCV NS5B replicase) palm site variant inhibitor with potency high oral bioavailability and pharmacokinetic parameters.BMS-929075 shows cytotoxicity. purity: 97%