Learn More
Selleck Chemical LLC BMS-935177 2mg 1231889-53-4
SDP
Supplier:  Selleck Chemical LLC S83482MG
Encompass_Preferred
BMS-935177 is a potent, reversible Bruton's Tyrosine Kinase (BTK) inhibitor with an IC50 value of 2.8 nM and demonstrates good kinase selectivity. It is more potent against BTK than other kinase, including the other Tec family kinases (TEC, BMX, ITK, and TXK) over which the compound is between 5- and 67-fold selective. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8304