Learn More
Medchemexpress LLC HY-101793 5mg Medchemexpress, BMS-935177 CAS:1231889-53-4 Purity:>98%
SDP
Catalog No. 501932219
Encompass_Preferred
Medchemexpress, HY-101793 5mg BMS-935177 CAS:1231889-53-4 BMS-935177 is a potent and selective reversible inhibitor of Bruton’s tyrosine kinase ( Btk ) with an IC 50 of 3 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2219 Supplier Medchemexpress LLC Supplier No. HY1017935MG
Medchemexpress, HY-101793 5mg BMS-935177 CAS:1231889-53-4 BMS-935177 is a potent and selective reversible inhibitor of Bruton’s tyrosine kinase ( Btk ) with an IC 50 of 3 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.