Learn More
Medchemexpress LLC BMS-962212 10mg | 1430114-34-3 | C32H28ClFN8O5 | 10 MG
SDP
Catalog No. 5000226247
Encompass_Preferred
BMS-962212 is a reversible, direct, and selective small-molecule inhibitor of coagulation factor XIa (FXIa) used in research on anticoagulation and thrombosis. It produces dose-dependent increases in activated partial thromboplastin time (aPTT) and reductions in FXI clotting activity; molecular formula C32H28ClFN8O5, molecular weight 659.07 g·mol⁻1.
  • Reversible, selective FXIa inhibition for anticoagulation research.
  • Demonstrated dose-dependent pharmacodynamic effects on aPTT and FXI activity.
  • High reported purity (99.33%).
  • Offered in research sample quantities (10 MG).
Catalog No. 50-002-26247 Supplier Medchemexpress LLC Supplier No. HY11729010MG
BMS-962212 is a reversible, direct, and selective small-molecule inhibitor of coagulation factor XIa (FXIa) used in research on anticoagulation and thrombosis. It produces dose-dependent increases in activated partial thromboplastin time (aPTT) and reductions in FXI clotting activity; molecular formula C32H28ClFN8O5, molecular weight 659.07 g·mol⁻1.
  • Reversible, selective FXIa inhibition for anticoagulation research.
  • Demonstrated dose-dependent pharmacodynamic effects on aPTT and FXI activity.
  • High reported purity (99.33%).
  • Offered in research sample quantities (10 MG).