Learn More
Medchemexpress LLC BMS-963272 | 1441057-15-3 | 99.0% | C24H21F6N5O2 | 10MG
SDP
Catalog No. 5000259646
Encompass_Preferred
BMS-963272 is an orally active, selective inhibitor of monoacylglycerol acyltransferase 2 (MGAT2) originally reported in the peer-reviewed medicinal chemistry literature. It exhibits potent MGAT2 inhibition (IC50 = 7.1 nM) and is used in preclinical research to probe MGAT2 biology and to study metabolic and inflammatory disease models.
  • Potent, selective MGAT2 inhibition (IC50 = 7.1 nM).
  • High purity suitable for research (≈99.0%).
  • Supplied as a solid powder with defined storage conditions for stability.
  • Applicable to in vitro and in vivo preclinical studies of metabolic and inflammatory disorders.
  • Structural identifiers available for cheminformatics use (SMILES, InChIKey).
Catalog No. 50-002-59646 Supplier Medchemexpress LLC Supplier No. HY13292410MG
BMS-963272 is an orally active, selective inhibitor of monoacylglycerol acyltransferase 2 (MGAT2) originally reported in the peer-reviewed medicinal chemistry literature. It exhibits potent MGAT2 inhibition (IC50 = 7.1 nM) and is used in preclinical research to probe MGAT2 biology and to study metabolic and inflammatory disease models.
  • Potent, selective MGAT2 inhibition (IC50 = 7.1 nM).
  • High purity suitable for research (≈99.0%).
  • Supplied as a solid powder with defined storage conditions for stability.
  • Applicable to in vitro and in vivo preclinical studies of metabolic and inflammatory disorders.
  • Structural identifiers available for cheminformatics use (SMILES, InChIKey).