Learn More
Selleck Chemical LLC BMS-986020 S3572-25mg
SDP
Catalog No. 503108965
Encompass_Preferred
BMS-986020 (AM152 AP-3152 free acid) is a potent and selective antagonist of lysophosphatidic acid receptor 1 (LPA1) BMS-986020 inhibits bile acid and phospholipid transporters with IC50 of 4 8 M 6 2 M and 7 5 M for BSEP MRP4 and MDR3 respectively BMS-986020 has the potential for the treatment of idiopathic pulmonary fibrosis (IPF)
Catalog No. 50-310-8965 Supplier Selleck Chemical LLC Supplier No. S357225MG
BMS-986020 (AM152 AP-3152 free acid) is a potent and selective antagonist of lysophosphatidic acid receptor 1 (LPA1) BMS-986020 inhibits bile acid and phospholipid transporters with IC50 of 4 8 M 6 2 M and 7 5 M for BSEP MRP4 and MDR3 respectively BMS-986020 has the potential for the treatment of idiopathic pulmonary fibrosis (IPF)