Learn More
Selleck Chemical LLC BMS-986020-5mg
SDP
Supplier:  Selleck Chemical LLC S35725MG
Encompass_Preferred
BMS-986020 (AM152, AP-3152 free acid) is a potent and selective antagonist of lysophosphatidic acid receptor 1 (LPA1). BMS-986020 inhibits bile acid and phospholipid transporters with IC50 of 4.8 μM, 6.2 μM, and 7.5 μM for BSEP, MRP4, and MDR3, respectively. BMS-986020 has the potential for the treatment of idiopathic pulmonary fibrosis (IPF).
Catalog No. 50-238-9609