Learn More
Abcam BMS-986020, 25MG
Supplier:  Abcam AB28698925MG
Encompass_Preferred
MW 482.5 Da, Purity >=98%. Potent G protein-coupled lysophosphatidic acid receptor 1 (LPA1) antagonist. LPA receptor is involved in binding of the signaling molecule lysophosphatidic acid, which in turn is involved in a host of diverse biological functions, such as cell proliferation, platelet aggregation, smooth muscle contraction and others.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-220-5857