Learn More
Medchemexpress LLC HY-100619A 10mg Medchemexpress, BMS-986020 (sodium) CAS:1380650-53-2 Purity:>98%
SDP
Catalog No. 501967759
Encompass_Preferred
Medchemexpress, HY-100619A 10mg BMS-986020 (sodium) CAS:1380650-53-2 BMS-986020 sodium is a high-affinity lysophosphatidic acid receptor 1 (LPA1) antagonist. BMS-986020 sodium inhibits bile acid and phospholipid transporters with IC50s of 4.8 µM, 6.2 µM, and 7.5 µM for BSEP, MRP4, and MDR3, respectively. BMS-986020 sodium has the potential for the treatment of idiopathic pulmonary fibrosis (IPF)[3]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-7759 Supplier Medchemexpress LLC Supplier No. HY100619A10MG
Medchemexpress, HY-100619A 10mg BMS-986020 (sodium) CAS:1380650-53-2 BMS-986020 sodium is a high-affinity lysophosphatidic acid receptor 1 (LPA1) antagonist. BMS-986020 sodium inhibits bile acid and phospholipid transporters with IC50s of 4.8 µM, 6.2 µM, and 7.5 µM for BSEP, MRP4, and MDR3, respectively. BMS-986020 sodium has the potential for the treatment of idiopathic pulmonary fibrosis (IPF)[3]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.