Learn More
Medchemexpress LLC Bms-986115 (notch inhibitor 1) | 1584647-27-7 | 99.9% | 10 MG
SDP
Catalog No. 5000341268
Encompass_Preferred
BMS-986115 is a potent Notch inhibitor used in cancer research.
  • Potent Notch inhibitor
  • Ic50s of 7.8 nM for Notch 1
  • Ic50s of 8.5 nM for Notch 3
Catalog No. 50-003-41268 Supplier Medchemexpress LLC Supplier No. HY1286010MG
BMS-986115 is a potent Notch inhibitor used in cancer research.
  • Potent Notch inhibitor
  • Ic50s of 7.8 nM for Notch 1
  • Ic50s of 8.5 nM for Notch 3