Learn More
Medchemexpress LLC BMS-986121 | 313671-26-0 | 100.0% | C15H9Cl2N3O2S | 25 MG
SDP
Catalog No. 5000352534
Encompass_Preferred
BMS-986121 is a positive allosteric modulator (PAM) of the μ opioid receptor, developed from patent WO2014107344. It is built on a novel chemical scaffold, representing a new chemotype for μ receptor PAMs.
  • Potent positive allosteric modulator of the μ opioid receptor.
  • Augments β-arrestin-recruitment response.
  • Increases inhibition of adenylyl cyclase activity.
  • Enhances the potency of endomorphin-I and leu-enkephalin in cAMP-accumulation assays.
Catalog No. 50-003-52534 Supplier Medchemexpress LLC Supplier No. 25MGHY141515
BMS-986121 is a positive allosteric modulator (PAM) of the μ opioid receptor, developed from patent WO2014107344. It is built on a novel chemical scaffold, representing a new chemotype for μ receptor PAMs.
  • Potent positive allosteric modulator of the μ opioid receptor.
  • Augments β-arrestin-recruitment response.
  • Increases inhibition of adenylyl cyclase activity.
  • Enhances the potency of endomorphin-I and leu-enkephalin in cAMP-accumulation assays.