Learn More
Medchemexpress LLC HY-101856 10mg Medchemexpress, BMS-986142 CAS:1643368-58-4 Purity:>98%
SDP
Catalog No. 501967891
Encompass_Preferred
Medchemexpress, HY-101856 10mg BMS-986142 CAS:1643368-58-4 BMS-986142 is a potent and highly selective reversible inhibitor of Bruton's tyrosine kinase (BTK) with an IC50 of 0.5 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-7891 Supplier Medchemexpress LLC Supplier No. HY10185610MG
Medchemexpress, HY-101856 10mg BMS-986142 CAS:1643368-58-4 BMS-986142 is a potent and highly selective reversible inhibitor of Bruton's tyrosine kinase (BTK) with an IC50 of 0.5 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.