Learn More
Abcam BMS-986142, 1MG
Catalog No. 502671214
Encompass_Preferred
MW 572.6 Da, Purity >=98%. BMS-986142 is a potent, selective, and reversible BTK (Bruton's tyrosine kinase) inhibitor (BTK IC₅₀ = 0.5nM; human WB IC₅₀ = 90 nM). BMS-986142 displays robust efficacy in murine models of rheumatoid arthritis (RA), including collagen-induced arthritis (CIA) and collagen antibody-induced arthritis (CAIA).
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-267-1214 Supplier Abcam Supplier No. AB2870441MG
MW 572.6 Da, Purity >=98%. BMS-986142 is a potent, selective, and reversible BTK (Bruton's tyrosine kinase) inhibitor (BTK IC₅₀ = 0.5nM; human WB IC₅₀ = 90 nM). BMS-986142 displays robust efficacy in murine models of rheumatoid arthritis (RA), including collagen-induced arthritis (CIA) and collagen antibody-induced arthritis (CAIA).
The product is subject to the following: Abcam Restricted Use Statement