Learn More
Selleck Chemical LLC BMS-986158-5mg
SDP
Catalog No. 502389903
Encompass_Preferred
BMS-986158 is a potent inhibitor of BET with IC50s of 6.6 nM and 5 nM in NCI-H211 small cell lung cancer (SCLC) cells and MDA-MB231 triple negative breast cancer (TNBC) cells, respectively.
Catalog No. 50-238-9903 Supplier Selleck Chemical LLC Supplier No. S96915MG
BMS-986158 is a potent inhibitor of BET with IC50s of 6.6 nM and 5 nM in NCI-H211 small cell lung cancer (SCLC) cells and MDA-MB231 triple negative breast cancer (TNBC) cells, respectively.