Learn More
Medchemexpress LLC HY-117287 10mg Medchemexpress, BMS-986165 CAS:1609392-27-9 Purity:>98%
SDP
Catalog No. 502028836
Encompass_Preferred
Medchemexpress, HY-117287 10mg BMS-986165 CAS:1609392-27-9 BMS-986165 is a highly selective, orally bioavailable allosteric TYK2 inhibitor for the treatment of autoimmune diseases, which selectively binds to TYK2 pseudokinase (JH2) domain (IC50=1.0 nM) and blocks receptor-mediated Tyk2 activation by stabilizing the regulatory JH2 domain. BMS-986165 inhibits IL-12/23 and type I IFN pathways[2]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-8836 Supplier Medchemexpress LLC Supplier No. HY11728710MG
Medchemexpress, HY-117287 10mg BMS-986165 CAS:1609392-27-9 BMS-986165 is a highly selective, orally bioavailable allosteric TYK2 inhibitor for the treatment of autoimmune diseases, which selectively binds to TYK2 pseudokinase (JH2) domain (IC50=1.0 nM) and blocks receptor-mediated Tyk2 activation by stabilizing the regulatory JH2 domain. BMS-986165 inhibits IL-12/23 and type I IFN pathways[2]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.