Learn More
Medchemexpress LLC BMS-986202 | 1771691-34-9 | 99.3% | C22H18D3FN6O3 | 10MG
SDP
Catalog No. 5000261050
Encompass_Preferred
BMS-986202 is a deuterated, orally active TYK2 inhibitor used in research to interrogate TYK2-mediated inflammatory signaling. It binds the TYK2 JH2 pseudokinase domain with very high potency (reported IC50 = 0.19 nM; Ki = 0.02 nM) and has been characterized in preclinical and clinical studies. Supplied as a high-purity research compound for biochemical and cell-based assays; not for human therapeutic use.
  • Deuterated analogue designed for improved metabolic stability.
  • High potency against TYK2 JH2, enabling sensitive biochemical assays.
  • Marked selectivity over related kinases, reducing off-target effects.
  • High purity (99.3%), supporting reproducible experimental results.
  • Available in multiple solid and solution pack sizes for assay flexibility.
  • Suitable for both in vitro and in vivo research applications.
  • Intended for research use only; not for clinical or human therapeutic use.
Catalog No. 50-002-61050 Supplier Medchemexpress LLC Supplier No. HY13196810MG
BMS-986202 is a deuterated, orally active TYK2 inhibitor used in research to interrogate TYK2-mediated inflammatory signaling. It binds the TYK2 JH2 pseudokinase domain with very high potency (reported IC50 = 0.19 nM; Ki = 0.02 nM) and has been characterized in preclinical and clinical studies. Supplied as a high-purity research compound for biochemical and cell-based assays; not for human therapeutic use.
  • Deuterated analogue designed for improved metabolic stability.
  • High potency against TYK2 JH2, enabling sensitive biochemical assays.
  • Marked selectivity over related kinases, reducing off-target effects.
  • High purity (99.3%), supporting reproducible experimental results.
  • Available in multiple solid and solution pack sizes for assay flexibility.
  • Suitable for both in vitro and in vivo research applications.
  • Intended for research use only; not for clinical or human therapeutic use.