Learn More
Medchemexpress LLC HY-139485 5mg , BMS-986224 CAS:2055200-88-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1394855MG
Encompass_Preferred
HY-139485 5mg BMS-986224 CAS: 2055200-88-7 BMS-986224 is a potent, selective and orally active APJ receptor agonist (Kd = 0.3 nM). BMS-986224 exhibits similar receptor binding and signaling profile to (Pyr1) apelin-13. BMS-986224 has the potential for the research of heart failure. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1824