Learn More
Medchemexpress LLC HY-139853 5mg , BMS-986278 CAS:2170126-74-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1398535MG
Encompass_Preferred
HY-139853 5mg BMS-986278 CAS: 2170126-74-4 BMS-986278 is a potent lysophosphatidic acid receptor 1 (LPA1) antagonist, with a human LPA1 Kb of 6.9 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0840