Learn More
Medchemexpress LLC BMS-986278 (admilparant) | 2170126-74-4 | 99.9% | C22H31N5O5 | 10MG
SDP
Catalog No. 5000260108
Encompass_Preferred
Admilparant is a potent, orally active lysophosphatidic acid receptor 1 (LPA1) antagonist used in preclinical research, notably for studies of pulmonary fibrotic diseases. It has been characterized for potency, formulation, and storage to support both in vitro and in vivo experiments.
  • Potent LPA1 antagonist with low-nanomolar activity (human Kb ≈ 6.9 nM, mouse ≈ 4.0 nM).
  • Suitable for in vitro and in vivo studies, with validated formulation options for dosing.
  • High purity supporting reproducible experimental results (reported ≈99.9% by batch HPLC).
  • Soluble in DMSO at high concentration for stock preparation (100 mg/mL).
  • Stable as a powdered solid under frozen storage for long-term use.
Catalog No. 50-002-60108 Supplier Medchemexpress LLC Supplier No. HY13985310MG
Add to Cart
Add to Cart
Admilparant is a potent, orally active lysophosphatidic acid receptor 1 (LPA1) antagonist used in preclinical research, notably for studies of pulmonary fibrotic diseases. It has been characterized for potency, formulation, and storage to support both in vitro and in vivo experiments.
  • Potent LPA1 antagonist with low-nanomolar activity (human Kb ≈ 6.9 nM, mouse ≈ 4.0 nM).
  • Suitable for in vitro and in vivo studies, with validated formulation options for dosing.
  • High purity supporting reproducible experimental results (reported ≈99.9% by batch HPLC).
  • Soluble in DMSO at high concentration for stock preparation (100 mg/mL).
  • Stable as a powdered solid under frozen storage for long-term use.