Learn More
Selleck Chemical LLC BMS-986278-E1036-5MG
SDP
Catalog No. 503138847
Encompass_Preferred
BMS-986278 is a potent and orally active antagonist of lysophosphatidic acid receptor 1 (LPA1) BMS-986278 exhibits Kbs of 6 9 nM and 4 0 nM for human and mouse LPA1 respectively and is widely used in the research of pulmonary fibrotic diseases
Catalog No. 50-313-8847 Supplier Selleck Chemical LLC Supplier No. E10365MG
BMS-986278 is a potent and orally active antagonist of lysophosphatidic acid receptor 1 (LPA1) BMS-986278 exhibits Kbs of 6 9 nM and 4 0 nM for human and mouse LPA1 respectively and is widely used in the research of pulmonary fibrotic diseases