Learn More
Selleck Chemical LLC BMS-986365 E1964-100MG
SDP
Catalog No. 5000039477
Encompass_Preferred
BMS-986365 (CC-94676) is a potent and selective heterobifunctional ligand-directed degrader (LDD) and antagonist of androgen receptor (AR) that effectively targets both wildtype and mutant forms of the androgen receptor (AR) in the cytoplasm and nucleus It rapidly degrades AR suppresses AR signaling and shows promising clinical activity in inhibiting tumor growth in patients with metastatic castration-resistant prostate cancer (mCRPC)
Catalog No. 50-000-39477 Supplier Selleck Chemical LLC Supplier No. E1964100MG
BMS-986365 (CC-94676) is a potent and selective heterobifunctional ligand-directed degrader (LDD) and antagonist of androgen receptor (AR) that effectively targets both wildtype and mutant forms of the androgen receptor (AR) in the cytoplasm and nucleus It rapidly degrades AR suppresses AR signaling and shows promising clinical activity in inhibiting tumor growth in patients with metastatic castration-resistant prostate cancer (mCRPC)