Learn More
Selleck Chemical LLC BMS-986397 E4725-25MG
SDP
Catalog No. 5000041955
Encompass_Preferred
BMS-986397 (CC-91633) is a potent and selective thalidomide-derived molecular glue degrader of CK1 and a modulator of CRBN E3 ubiquitin ligase As a first-in-class CELMoD agent it exhibits strong anti-leukemic activity in AML (Acute Myeloid Leukemia) and HR-MDS (High-Risk Myelodysplastic Syndrome) models with enhanced efficacy and potency for hematologic malignancies and immunomodulation
Catalog No. 50-000-41955 Supplier Selleck Chemical LLC Supplier No. E472525MG
BMS-986397 (CC-91633) is a potent and selective thalidomide-derived molecular glue degrader of CK1 and a modulator of CRBN E3 ubiquitin ligase As a first-in-class CELMoD agent it exhibits strong anti-leukemic activity in AML (Acute Myeloid Leukemia) and HR-MDS (High-Risk Myelodysplastic Syndrome) models with enhanced efficacy and potency for hematologic malignancies and immunomodulation